Anti-MLX Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MLX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-226
- 77859-830
- Antigen Symbol:
- MLX
- MLX
- Antigen Synonyms:
- MLXIP|TCFL4|Transcription factor-like protein 4|MAD7|Max-like bHLHZip protein|MONDOA|MLX_HUMAN|transcription factor like 4|Transcription factor like protein 4|Max like protein X|bHLHd13|Max like bHLHZip protein|BigMax protein|Class D basic helix-loop-helix protein 13|MXD7|Protein BigMax|Protein Max-like protein X
- bHLHd13|BigMax protein|Class D basic helix-loop-helix protein 13|MAD7|Max like bHLHZip protein|Max like protein X|Max-like bHLHZip protein|Max-like protein X|MLX_HUMAN|MLXIP|MONDOA|MXD7|Protein BigMax|Protein TCFL4|transcription factor like 4|Transcription factor like protein 4|Transcription factor-like protein 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UH92
- Q9UH92
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Max-like protein X protein (1-244AA)
- Recombinant Human Max-like protein X protein (1-244AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together