Anti-GBP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: GBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-833
- 77738-029
- Antigen Symbol:
- GBP1
- GBP1
- Antigen Synonyms:
- GBP 1|GBP-1|GBP1_HUMAN|GTP binding protein 1|GTP-binding protein 1|Guanine nucleotide binding protein 1|Guanine nucleotide-binding protein 1|Guanylate binding protein 1|Guanylate binding protein 1 interferon inducible 67kDa|Guanylate binding protein 1 interferon inducible|HuGBP 1|HuGBP-1|HuInterferon induced guanylate binding protein 1|Interferon-induced guanylate-binding protein 1|OTTHUMP00000012352
- GBP 1|GBP-1|GBP1_HUMAN|GTP binding protein 1|GTP-binding protein 1|Guanine nucleotide binding protein 1|Guanine nucleotide-binding protein 1|Guanylate binding protein 1|Guanylate binding protein 1 interferon inducible 67kDa|Guanylate binding protein 1 interferon inducible|HuGBP 1|HuGBP-1|HuInterferon induced guanylate binding protein 1|Interferon-induced guanylate-binding protein 1|OTTHUMP00000012352
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32455
- P32455
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon-induced guanylate-binding protein 1 protein (383-592AA)
- Recombinant Human Interferon-induced guanylate-binding protein 1 protein (383-592AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together