Anti-HS3ST1 Rabbit Polyclonal Antibody
Catalog # 77740-396
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HS3ST1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O14792
- Antigen Synonyms:3-OST-1|3OST|3OST1|EC 2.8.2.23|h3 OST 1|h3-OST-1|Heparan sulfate (glucosamine) 3 O sulfotransferase 1|Heparan sulfate 3 O sulfotransferase 1|Heparan sulfate 3-O-sulfotransferase 1|Heparan sulfate D glucosaminyl 3 O sulfotransferase 1|Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1|Heparan sulfate glucosamine 3-O-sulfotransferase 1|Heparin glucosamine 3 O sulfotransferase|HS3S1_HUMAN|HS3ST 1|Hs3st1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:3-OST-1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HS3ST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat