Anti-NTHL1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NTHL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-077
- 77745-076
- Antigen Symbol:
- NTHL1
- NTHL1
- Antigen Synonyms:
- Bifunctional DNA N glycoslyase/DNA (apurinic or apyrimidinic site) lyase|DNA glycoslyase/AP lyase|Endonuclease III like protein 1|Endonuclease III-like protein 1|hNTH1|NTH 1|nth endonuclease III like 1 (E. coli)|NTH endonuclease III Like 1|NTH1|NTHL 1|NTHL1_HUMAN|OCTS 3|OCTS3
- Bifunctional DNA N glycoslyase/DNA (apurinic or apyrimidinic site) lyase|DNA glycoslyase/AP lyase|Endonuclease III like protein 1|Endonuclease III-like protein 1|hNTH1|NTH 1|nth endonuclease III like 1 (E. coli)|NTH endonuclease III Like 1|NTH1|NTHL 1|NTHL1_HUMAN|OCTS 3|OCTS3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78549
- P78549
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Endonuclease III-like protein 1 protein (31-312AA)
- Endonuclease III-like protein 1 protein (31-312AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together