Anti-HMBS Rabbit Polyclonal Antibody
77740-130
New Product
- Antigen Symbol:HMBS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P08397
- Antigen Synonyms:acute|Uroporphyrinogen I synthase|PBG-D|PBGD|Uroporphyrinogen I synthetase|Hydroxymethylbilane synthase|Chester type|PORC|porphyria|Pre-uroporphyrinogen synthase|UPS|Pre uroporphyrinogen synthase|HEM3_HUMAN|Porphobilinogen deaminase|PBG D
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Uroporphyrinogen I synthetase
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HMBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat