Anti-GNAL Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNAL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-533
- 77856-170
- Antigen Symbol:
- GNAL
- GNAL
- Antigen Synonyms:
- alpha stimulating activity polypeptide|DYT25|GNAL_HUMAN|alpha activating activity polypeptide|olfactory type|Adenylate cyclase-stimulating G alpha protein|Guanine nucleotide binding protein (G protein)|Guanine nucleotide binding protein G(olf) subunit alpha|Guanine nucleotide-binding protein G(olf) subunit alpha|Adenylate cyclase stimulating G alpha protein|G alpha olf
- Adenylate cyclase stimulating G alpha protein|olfactory type|Adenylate cyclase-stimulating G alpha protein|DYT25|G alpha olf|GNAL_HUMAN|Guanine nucleotide binding protein (G protein)|alpha activating activity polypeptide|alpha stimulating activity polypeptide|Guanine nucleotide binding protein G(olf) subunit alpha|Guanine nucleotide-binding protein G(olf) subunit alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P38405
- P38405
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein G(olf) subunit alpha protein (4-116AA)
- Recombinant Human Guanine nucleotide-binding protein G(olf) subunit alpha protein (4-116AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together