Anti-HTRA2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HTRA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-533
- 77857-767
- Antigen Symbol:
- HTRA2
- Antigen Synonyms:
- High temperature requirement protein A2|HTRA 2|HtrA like serine protease|HtrA serine peptidase 2|HtrA|E. coli|homolog of|2|HtrA2|HTRA2_HUMAN|mitochondrial|Omi stress regulated endoprotease|Omi stress-regulated endoprotease|PARK 13|PARK13|Protease serine 25|PRSS 25|PRSS25|Serine protease 25|Serine protease HTRA2|Serine protease HTRA2 mitochondrial|Serine protease htra2 mitochondrial precursor|Serine protease omi|Serine proteinase OMI
- High temperature requirement protein A2|HTRA 2|HtrA like serine protease|HtrA serine peptidase 2|HtrA|E. coli|homolog of|2|HtrA2|HTRA2_HUMAN|mitochondrial|Omi stress regulated endoprotease|Omi stress-regulated endoprotease|PARK 13|PARK13|Protease serine 25|PRSS 25|PRSS25|Serine protease 25|Serine protease HTRA2|Serine protease HTRA2 mitochondrial|Serine protease htra2 mitochondrial precursor|Serine protease omi|Serine proteinase OMI
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O43464
- O43464
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Serine protease HTRA2, mitochondrial protein (334-458AA)
- Recombinant Human Serine protease HTRA2, mitochondrial protein (334-458AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together