Anti-MMP3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:500 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: MMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-174
- 77859-793
- Antigen Symbol:
- MMP3
- MMP3
- Antigen Synonyms:
- CHDS6|Matrix metalloproteinase 3|Matrix metalloproteinase 3 preproprotein|Matrix metalloproteinase-3|MGC126102|MGC126103|MGC126104|MMP 3|MMP-3|MMP3_HUMAN|Proteoglycanase|SL-1|SL1|STMY|STMY1|STR1|Stromelisin 1|Stromelysin 1|Stromelysin 1 progelatinase|Stromelysin-1|Transin 1|Transin-1
- CHDS6|Matrix metalloproteinase 3|Matrix metalloproteinase 3 preproprotein|Matrix metalloproteinase-3|MGC126102|MGC126103|MGC126104|MMP 3|MMP-3|MMP3_HUMAN|Proteoglycanase|SL-1|SL1|STMY|STMY1|STR1|Stromelisin 1|Stromelysin 1|Stromelysin 1 progelatinase|Stromelysin-1|Transin 1|Transin-1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08254
- P08254
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MMP3
- Fusion protein of Human MMP3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together