Anti-RACK1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RACK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-573
- 77856-205
- Antigen Symbol:
- RACK1
- Antigen Synonyms:
- Cell proliferation-inducing gene 21 protein|GBLP_HUMAN|Gnb2-rs1|Gnb2l1|Guanine nucleotide binding protein (G protein) beta polypeptide 2 like 1|Guanine nucleotide binding protein beta polypeptide 2 like 1|Guanine nucleotide binding protein beta subunit 2 like 1|Guanine nucleotide binding protein beta subunit like protein 12.3|Guanine nucleotide binding protein subunit beta 2 like 1|Guanine nucleotide binding protein subunit beta like protein 12.3|Guanine nucleotide-binding protein subunit beta-2-like 1|Guanine nucleotide-binding protein subunit beta-like protein 12.3|H12.3|HLC-7|Human lung cancer oncogene 7 protein|lung cancer oncogene 7|OTTHUMP00000223704|OTTHUMP00000223870|OTTHUMP00000223891|OTTHUMP00000223893|OTTHUMP00000223900|OTTHUMP00000223902|OTTHUMP00000223930|OTTHUMP00000223931|PIG21|Proliferation inducing gene 21|Protein homologous to chicken B complex protein guanine nucleotide binding|RACK1|Receptor for activated C kinase 1|Receptor for activated C kinase|Receptor of activated protein kinase C 1
- Cell proliferation-inducing gene 21 protein|GBLP_HUMAN|Gnb2-rs1|Gnb2l1|Guanine nucleotide binding protein (G protein) beta polypeptide 2 like 1|Guanine nucleotide binding protein beta polypeptide 2 like 1|Guanine nucleotide binding protein beta subunit 2 like 1|Guanine nucleotide binding protein beta subunit like protein 12.3|Guanine nucleotide binding protein subunit beta 2 like 1|Guanine nucleotide binding protein subunit beta like protein 12.3|Guanine nucleotide-binding protein subunit beta-2-like 1|Guanine nucleotide-binding protein subunit beta-like protein 12.3|H12.3|HLC-7|Human lung cancer oncogene 7 protein|lung cancer oncogene 7|OTTHUMP00000223704|OTTHUMP00000223870|OTTHUMP00000223891|OTTHUMP00000223893|OTTHUMP00000223900|OTTHUMP00000223902|OTTHUMP00000223930|OTTHUMP00000223931|PIG21|Proliferation inducing gene 21|Protein homologous to chicken B complex protein guanine nucleotide binding|RACK1|Receptor for activated C kinase 1|Receptor for activated C kinase|Receptor of activated protein kinase C 1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P63244
- P63244
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Receptor of activated protein C kinase 1 protein (93-317AA)
- Recombinant Human Receptor of activated protein C kinase 1 protein (93-317AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together