Anti-KCNJ1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNJ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-435
- 77741-487
- Antigen Symbol:
- KCNJ1
- KCNJ1
- Antigen Synonyms:
- ATP regulated potassium channel ROM K|ATP sensitive inward rectifier potassium channel 1|ATP-regulated potassium channel ROM-K|ATP-sensitive inward rectifier potassium channel 1|Inward rectifier K(+) channel Kir1.1|inwardly rectifying K+ channel|inwardly rectifying subfamily J member 1|IRK1_HUMAN|KCNJ 1|KCNJ|Kir 1.1|Kir1.1|OTTHUMP00000045938|Potassium channel|Potassium channel inwardly rectifying subfamily J member 1|potassium inwardly-rectifying channel J1|ROMK 1|ROMK 2|ROMK|ROMK1|ROMK2
- Potassium channel inwardly rectifying subfamily J member 1|inwardly rectifying subfamily J member 1|ROMK 1|ROMK 2|ATP-regulated potassium channel ROM-K|potassium inwardly-rectifying channel J1|Inward rectifier K(+) channel Kir1.1|ATP regulated potassium channel ROM K|KCNJ 1|ATP sensitive inward rectifier potassium channel 1|ATP-sensitive inward rectifier potassium channel 1|KCNJ|Kir1.1|inwardly rectifying K+ channel|IRK1_HUMAN|OTTHUMP00000045938|Kir 1.1|Potassium channel|ROMK|ROMK1|ROMK2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48048
- P48048
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human ATP-sensitive inward rectifier potassium channel 1 protein (178-391AA)
- Recombinant Human ATP-sensitive inward rectifier potassium channel 1 protein (178-391AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together