Anti-MSI2 Rabbit Polyclonal Antibody
Catalog # 77743-523
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MSI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96DH6
- Antigen Synonyms:FLJ36569|MGC3245|Msi2|MSI2/HOXA9 fusion gene|included|MSI2H|MSI2H_HUMAN|Musashi 2|Musashi homolog 2|Musashi RNA binding protein 2|Musashi|Drosophila|homolog of|2|Musashi-2|RNA binding protein Musashi homolog 2|RNA-binding protein Musashi homolog 2|WD 40 repeat protein MSI2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Musashi-2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MSI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat