Anti-ACTN4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACTN4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-637
- 77855-156
- Antigen Symbol:
- ACTN4
- ACTN4
- Antigen Synonyms:
- actinin 4|Actinin alpha 4|actinin4|ACTN 4|ACTN4_HUMAN|Alpha-actinin-4|DKFZp686K23158|F actin cross linking protein|F-actin cross-linking protein|Focal segmental glomerulosclerosis 1|FSGS 1|FSGS|FSGS1|Non muscle alpha actinin 4|Non-muscle alpha-actinin 4
- actinin 4|Actinin alpha 4|actinin4|ACTN 4|ACTN4_HUMAN|Alpha-actinin-4|DKFZp686K23158|F actin cross linking protein|F-actin cross-linking protein|Focal segmental glomerulosclerosis 1|FSGS 1|FSGS|FSGS1|Non muscle alpha actinin 4|Non-muscle alpha-actinin 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43707
- O43707
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Alpha-actinin-4 protein (6-277AA)
- Recombinant human Alpha-actinin-4 protein (6-277AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together