Anti-HOMER1 Rabbit Polyclonal Antibody
77740-263
New Product
- Antigen Symbol:HOMER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q86YM7
- Antigen Synonyms:HOME1_HUMAN|Homer|HOMER 1A|HOMER 1B|HOMER 1C|Homer homolog 1 (Drosophila)|homer homolog 1|Homer protein homolog 1|homer scaffolding protein 1|Drosophila|homolog of|1|neuronal immediate early gene|Homer-1|Homer1|HOMER1A|HOMER1B|HOMER1C|SYN47|Ves 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ves 1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HOMER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat