Anti-ID4 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50-1:200
Type:
Antigen: ID4
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-787
- 77740-571
- Antigen Symbol:
- ID4
- ID4
- Antigen Synonyms:
- bHLHb27|Class B basic helix-loop-helix protein 27|DNA binding protein inhibitor ID 4|DNA binding protein inhibitor DNA-binding protein inhibitor ID-4|ID 4|ID4_HUMAN|IDB4|Inhibitor of DNA binding 4|Inhibitor of DNA binding 4 dominant negative helix loop helix protein
- bHLHb27| Class B basic helix-loop-helix protein 27| DNA binding protein inhibitor ID 4| DNA binding protein inhibitor DNA-binding protein inhibitor ID-4| ID 4| ID4_HUMAN| IDB4| Inhibitor of DNA binding 4| Inhibitor of DNA binding 4 dominant negative helix loop helix protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47928
- P47928
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA-binding protein inhibitor ID-4 protein (1-161AA)
- Recombinant Human DNA-binding protein inhibitor ID-4 protein (1-161AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together