Anti-HTRA1 Rabbit Polyclonal Antibody
77740-531
New Product
- Antigen Symbol:HTRA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q92743
- Antigen Synonyms:ARMD7|CARASIL|High-temperature requirement A serine peptidase 1|HtrA|HtrA serine peptidase 1|HTRA1|HTRA1_HUMAN|IGFBP5 protease|L56|ORF480|Protease serine 11 (IGF binding)|protease serine 11|PRSS11|Serine protease 11|Serine protease HTRA1|Serine protease HTRA1 precursor
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Serine protease HTRA1 precursor
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HTRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat