Anti-NARS Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NARS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-930
- 77743-929
- Antigen Symbol:
- NARS
- NARS
- Antigen Synonyms:
- 3010001M15Rik|AA960128|AsnRS|ASNS|Asparagine tRNA ligase 1|cytoplasmic|Asparagine tRNA ligase|Asparagine--tRNA ligase|Asparaginyl tRNA synthetase|Asparaginyl-tRNA synthetase|C78150|EC 6.1.1.22|LRRGT00113|MGC116236|NARS1|NRS|SYNC_HUMAN
- 3010001M15Rik|AA960128|AsnRS|ASNS|Asparagine tRNA ligase 1|cytoplasmic|Asparagine tRNA ligase|Asparagine--tRNA ligase|Asparaginyl tRNA synthetase|Asparaginyl-tRNA synthetase|C78150|EC 6.1.1.22|LRRGT00113|MGC116236|NARS1|NRS|SYNC_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43776
- O43776
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Asparagine-tRNA ligase, cytoplasmic protein (1-300AA)
- Asparagine-tRNA ligase, cytoplasmic protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together