Anti-IRX3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IRX3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-225
- 77858-227
- Antigen Symbol:
- IRX3
- IRX3
- Antigen Synonyms:
- AI894186|Homeodomain protein IRXB1|Iroquois class homeodomain protein IRX 1|Iroquois homeobox 3|Iroquois homeobox protein 3|Iroquois-class homeodomain protein irx-3|IRX 1|IRX3_HUMAN|IRXB1
- AI894186|Homeodomain protein IRXB1|Iroquois class homeodomain protein IRX 1|Iroquois homeobox 3|Iroquois homeobox protein 3|Iroquois-class homeodomain protein irx-3|IRX 1|IRX3_HUMAN|IRXB1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78415
- P78415
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Iroquois-class homeodomain protein IRX-3 protein (285-378AA)
- Recombinant Human Iroquois-class homeodomain protein IRX-3 protein (285-378AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together