Anti-NCOR1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NCOR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-070
- 77744-069
- Antigen Symbol:
- NCOR1
- NCOR1
- Antigen Synonyms:
- hN CoR|hNCoR|KIAA1047|N CoR|N Cor/SMRT corepressor Rip13|N CoR1|N-CoR|N-CoR1|NCOR 1|NCoR|Ncor1|NCOR1_HUMAN|Nuclear receptor co repressor 1|Nuclear receptor corepressor 1|Retinoid X receptor interacting protein 13|RIP13|Rxrip13|thyroid hormone and retinoic acid receptor associated corepressor 1|TRAC 1|TRAC1
- hN CoR|hNCoR|KIAA1047|N CoR|N Cor/SMRT corepressor Rip13|N CoR1|N-CoR|N-CoR1|NCOR 1|NCoR|Ncor1|NCOR1_HUMAN|Nuclear receptor co repressor 1|Nuclear receptor corepressor 1|Retinoid X receptor interacting protein 13|RIP13|Rxrip13|thyroid hormone and retinoic acid receptor associated corepressor 1|TRAC 1|TRAC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75376
- O75376
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear receptor corepressor 1 protein (1770-1947AA)
- Nuclear receptor corepressor 1 protein (1770-1947AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together