Anti-JCHAIN Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: JCHAIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-961
- 77740-878
- Antigen Symbol:
- JCHAIN
- JCHAIN
- Antigen Synonyms:
- IGCJ|IGJ_HUMAN|Immunoglobulin J chain|Immunoglobulin J polypeptide linker protein for immunoglobulin alpha and mu polypeptides|immunoglobulin joining chain|JCH|Joining chain of multimeric IgA and IgM
- IGCJ|IGJ_HUMAN|Immunoglobulin J chain|Immunoglobulin J polypeptide linker protein for immunoglobulin alpha and mu polypeptides|immunoglobulin joining chain|JCH|Joining chain of multimeric IgA and IgM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01591
- P01591
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Immunoglobulin J chain protein (23-159AA)
- Recombinant human Immunoglobulin J chain protein (23-159AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together