Anti-KCNJ4 Rabbit Polyclonal Antibody
Catalog # 77740-116
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KCNJ4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P48050
- Antigen Synonyms:Hippocampal inward rectifier|Hippocampal inward rectifier potassium channel|HIR|HIRK2|HRK1|Inward rectifier K(+) channel Kir2.3|Inward rectifier K+ channel Kir2.3|Inward rectifier potassium channel 4|Inward rectifier potassium channel Kir 2.3|inwardly rectifying subfamily J member 4|IRK-3|IRK3|IRK4_HUMAN|KCNJ 4|Kir2.3|MGC142066|MGC142068|OTTHUMP00000028922|Potassium channel|Potassium channel inwardly rectifying subfamily J member 4|Potassium inwardly rectifying channel J4|Potassium inwardly rectifying channel subfamily J member 4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Hippocampal inward rectifier
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:40000
Type:
Antigen: KCNJ4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat