Anti-MTHFD2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MTHFD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-586
- 77860-084
- Antigen Symbol:
- MTHFD2
- MTHFD2
- Antigen Synonyms:
- Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase|mitochondrial|Descriptions|Methenyltetrahydrofolate cyclohydrolase|methylene tetrahydrofolate dehydrogenase (NAD+ dependent)|methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2|methylenetetrahydrofolate dehydrogenase (NADP+ dependent)|methylenetetrahydrofolate dehydrogenase 2|MGC82516|MTDC_HUMAN|mthfd2 methylene tetrahydrofolate dehydrogenase (NAD+ dependent)|NAD-dependent methylene tetrahydrofolate dehydrogenase|NAD-dependent methylenetetrahydrofolate dehydrogenase|NMDMC
- Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase|mitochondrial|Descriptions|Methenyltetrahydrofolate cyclohydrolase|methylene tetrahydrofolate dehydrogenase (NAD+ dependent)|methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2|methylenetetrahydrofolate dehydrogenase (NADP+ dependent)|methylenetetrahydrofolate dehydrogenase 2|MGC82516|MTDC_HUMAN|mthfd2 methylene tetrahydrofolate dehydrogenase (NAD+ dependent)|NAD-dependent methylene tetrahydrofolate dehydrogenase|NAD-dependent methylenetetrahydrofolate dehydrogenase|NMDMC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13995
- P13995
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial protein (40-250AA)
- Recombinant Human Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial protein (40-250AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together