Anti-GC Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: GC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77855-846
- 77738-048
- Antigen Symbol:
- GC
- GC
- Antigen Synonyms:
- DBP|DBP/Gc globulin|Gc-globulin|GRD3|Group specific component|Group specific component vitamin D binding protein|Group-specific component|hDBP|VDB|VDBG|VDBP|Vitamin D binding alpha globulin|Vitamin D-binding protein|VTDB_HUMAN
- DBP|DBP/Gc globulin|Gc-globulin|GRD3|Group specific component|Group specific component vitamin D binding protein|Group-specific component|hDBP|VDB|VDBG|VDBP|Vitamin D binding alpha globulin|Vitamin D-binding protein|VTDB_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02774
- P02774
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Vitamin D-binding protein (305-474AA)
- Recombinant human Vitamin D-binding protein (305-474AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together