Anti-MRTO4 Rabbit Polyclonal Antibody
Catalog # 77743-488
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRTO4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UKD2
- Antigen Synonyms:C1orf33|MRT4|mRNA turnover protein 4 homolog|Ribosome assembly factor MRTO4
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ribosome assembly factor MRTO4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MRTO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat