Anti-FKBP7 Rabbit Polyclonal Antibody
77737-252
New Product
- Antigen Symbol:FKBP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y680
- Antigen Synonyms:FKBP23|UNQ670/PRO1304Peptidyl-prolyl cis-trans isomerase PPIase EC 5.2.1.8|23 kDa FK506-binding protein|23 kDa FKBP|FKBP-23|FK506-binding protein 7|FKBP-7|Rotamase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Rotamase
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: FKBP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat