Anti-MFN2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MFN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-073
- 77859-712
- Antigen Symbol:
- MFN2
- MFN2
- Antigen Synonyms:
- CMT2A|CMT2A2|CPRP 1|CPRP1|EC 3.6.5.-|Fzo|HSG|hyperplasia suppressor gene|Hypertension related protein 1|KIAA0214|MARF|MFN 2|MFN2_HUMAN|Mitochondrial assembly regulatory factor|Mitofusin-2|Mitofusin2|Transmembrane GTPase MFN2
- CMT2A|CMT2A2|CPRP 1|CPRP1|EC 3.6.5.-|Fzo|HSG|hyperplasia suppressor gene|Hypertension related protein 1|KIAA0214|MARF|MFN 2|MFN2_HUMAN|Mitochondrial assembly regulatory factor|Mitofusin-2|Mitofusin2|Transmembrane GTPase MFN2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95140
- O95140
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitofusin-2 protein (61-158AA)
- Recombinant Human Mitofusin-2 protein (61-158AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together