Anti-LYRM1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LYRM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-279
- 77742-517
- Antigen Symbol:
- LYRM1
- LYRM1
- Antigen Synonyms:
- 1110065L10Rik|2310004B22Rik|4930404J24Rik|A211C6.1|LYR motif containing 1|LYR motif containing protein 1|LYR motif-containing protein 1|LYRM 1|lyrm1|LYRM1_HUMAN|OTTHUMP00000162292|OTTMUSP00000031126|OTTMUSP00000031127|OTTMUSP00000031128|OTTMUSP00000031129|RGD1563498
- 1110065L10Rik|2310004B22Rik|4930404J24Rik|A211C6.1|LYR motif containing 1|LYR motif containing protein 1|LYR motif-containing protein 1|LYRM 1|lyrm1|LYRM1_HUMAN|OTTHUMP00000162292|OTTMUSP00000031126|OTTMUSP00000031127|OTTMUSP00000031128|OTTMUSP00000031129|RGD1563498
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43325
- O43325
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human LYR motif-containing protein 1 protein (1-122AA)
- Recombinant Human LYR motif-containing protein 1 protein (1-122AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together