Anti-MRPS36 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPS36
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-480
- 77859-998
- Antigen Symbol:
- MRPS36
- MRPS36
- Antigen Synonyms:
- 1110018B13Rik|28S ribosomal protein S36|DC47|MGC22896|mitochondrial|MRP-S36|OTTHUMP00000222036|RT36_HUMAN|S36mt
- 1110018B13Rik|28S ribosomal protein S36|DC47|MGC22896|mitochondrial|MRP-S36|OTTHUMP00000222036|RT36_HUMAN|S36mt
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P82909
- P82909
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 28S ribosomal protein S36, mitochondrial protein (1-103AA)
- Recombinant Human 28S ribosomal protein S36, mitochondrial protein (1-103AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together