Anti-FMNL1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: FMNL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-316
- 77855-486
- Antigen Symbol:
- FMNL1
- FMNL1
- Antigen Synonyms:
- 8030453N10Rik|AI553564|C17orf1|C17orf1B|CLL associated antigen KW 13|CLL-associated antigen KW-13|FHOD4|FMNL|FMNL_HUMAN|Fnrl|Formin like 1|Formin like|Formin related protein|Formin-like protein 1|Frls|KW 13|KW13|Leukocyte formin|MGC133052|MGC1894|MGC21878
- 8030453N10Rik|AI553564|C17orf1|C17orf1B|CLL associated antigen KW 13|CLL-associated antigen KW-13|FHOD4|FMNL|FMNL_HUMAN|Fnrl|Formin like 1|Formin like|Formin related protein|Formin-like protein 1|Frls|KW 13|KW13|Leukocyte formin|MGC133052|MGC1894|MGC21878
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95466
- O95466
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human formin-like protein 1 protein (207-457AA)
- Recombinant human Formin-like protein 1 protein (207-457AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together