Anti-MAP3K4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MAP3K4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-671
- 77859-396
- Antigen Symbol:
- MAP3K4
- MAP3K4
- Antigen Synonyms:
- dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4)|FLJ42439|KIAA0213|M3K4_HUMAN|MAP three kinase 1|MAP/ERK kinase kinase 4|MAPK/ERK kinase kinase 4|MAPKKK4|MEK kinase 4|MEKK 4|MEKK4|Mitogen-activated protein kinase kinase kinase 4|MTK1|predicted protein of HQ0412|PRO0412|RP3-473J16.4|SSK2/SSK22 MAP kinase kinase kinase|yeast|homolog of
- dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4)|FLJ42439|KIAA0213|M3K4_HUMAN|MAP three kinase 1|MAP/ERK kinase kinase 4|MAPK/ERK kinase kinase 4|MAPKKK4|MEK kinase 4|MEKK 4|MEKK4|Mitogen-activated protein kinase kinase kinase 4|MTK1|predicted protein of HQ0412|PRO0412|RP3-473J16.4|SSK2/SSK22 MAP kinase kinase kinase|yeast|homolog of
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6R4
- Q9Y6R4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Mitogen-activated protein kinase kinase kinase 4 protein (53-145AA)
- Recombinant human Mitogen-activated protein kinase kinase kinase 4 protein (53-145AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together