Anti-HPGD Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: HPGD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77740-351
- 77857-627
- Antigen Symbol:
- HPGD
- HPGD
- Antigen Synonyms:
- 15 hydroxyprostaglandin dehydrogenase [NAD+]|15 PGDH|15-hydroxyprostaglandin dehydrogenase [NAD+]|15-PGDH|15PGDH|Hydroxyprostaglandin dehydrogenase 15 (NAD)|NAD+ dependent 15 hydroxyprostaglandin dehydrogenase|OTTHUMP00000218960|OTTHUMP00000219016|OTTHUMP00000219018|PGDH|PGDH_HUMAN|PGDH1|PHOAR1|Prostaglandin dehydrogenase 1|SDR36C1|Short chain dehydrogenase/reductase family 36C member 1
- 15 hydroxyprostaglandin dehydrogenase [NAD+]|15 PGDH|15-hydroxyprostaglandin dehydrogenase [NAD+]|15-PGDH|15PGDH|Hydroxyprostaglandin dehydrogenase 15 (NAD)|NAD+ dependent 15 hydroxyprostaglandin dehydrogenase|OTTHUMP00000218960|OTTHUMP00000219016|OTTHUMP00000219018|PGDH|PGDH_HUMAN|PGDH1|PHOAR1|Prostaglandin dehydrogenase 1|SDR36C1|Short chain dehydrogenase/reductase family 36C member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15428
- P15428
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 15-hydroxyprostaglandin dehydrogenase [NAD(+)] protein (14-130AA)
- Recombinant Human 15-hydroxyprostaglandin dehydrogenase [NAD(+)] protein (14-130AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at â20 or â80 °C. Avoid repeated freeze.
- Upon receipt, store at â20 or â80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together