Anti-ELK1 Rabbit Polyclonal Antibody
77735-811
New Product
- Antigen Symbol:ELK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P19419
- Antigen Synonyms:ELK 1|ELK1 member of ETS oncogene family|ELK1 protein|ETS transcription factor|ELK1_HUMAN|ELK2 member of ETS oncogene family|ETS domain containing protein Elk 1|ETS domain containing protein ETS domain protein ETS domain-containing protein Elk-1|ETS like gene 1|Member of ETS oncogene family|Oncogene Tyrosine kinase (ELK1) oncogene
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Tyrosine kinase (ELK1) oncogene
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: ELK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat