Anti-MRPL33 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL33
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-962
- 77743-413
- Antigen Symbol:
- MRPL33
- MRPL33
- Antigen Synonyms:
- 39S ribosomal protein L33|L33mt|mitochondrial|MRP-L33|RM33_HUMAN
- 39S ribosomal protein L33| L33mt| mitochondrial| MRP-L33| RM33_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75394
- O75394
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 39S ribosomal protein L33, mitochondrial protein (1-65AA)
- Recombinant Human 39S ribosomal protein L33, mitochondrial protein (1-65AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together