Anti-MTHFR Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: MTHFR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77860-089
- 77743-591
- Antigen Symbol:
- MTHFR
- MTHFR
- Antigen Synonyms:
- 5 10 methylenetetrahydrofolate reductase (NADPH)|5 10 methylenetetrahydrofolate reductase|Methylenetetrahydrofolate reductase (NAD(P)H)|Methylenetetrahydrofolate reductase|Methylenetetrahydrofolate reductase intermediate form|MTHFR|MTHR_HUMAN
- 5 10 methylenetetrahydrofolate reductase (NADPH)|5 10 methylenetetrahydrofolate reductase|Methylenetetrahydrofolate reductase (NAD(P)H)|Methylenetetrahydrofolate reductase|Methylenetetrahydrofolate reductase intermediate form|MTHFR|MTHR_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42898
- P42898
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Methylenetetrahydrofolate reductase protein (1-656AA)
- Recombinant Human Methylenetetrahydrofolate reductase protein (1-656AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together