Anti-INTS3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INTS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-143
- 77741-128
- Antigen Symbol:
- INTS3
- INTS3
- Antigen Synonyms:
- C1orf193|C1orf60|Integrator complex subunit 3|Int3|SOSS complex subunit A|Sensor of single-strand DNA complex subunit A|SOSS-A|Sensor of ssDNA subunit A
- C1orf193|C1orf60|Integrator complex subunit 3|Int3|SOSS complex subunit A|Sensor of single-strand DNA complex subunit A|SOSS-A|Sensor of ssDNA subunit A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q68E01
- Q68E01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integrator complex subunit 3 protein (901-1043AA)
- Recombinant Human Integrator complex subunit 3 protein (901-1043AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together