Anti-NPPB Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: NPPB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77744-826
- 77744-825
- Antigen Symbol:
- NPPB
- NPPB
- Antigen Synonyms:
- ANFB_HUMAN|BNP|BNP(1-32)|BNP(5-29)|BNP-32|Brain type natriuretic peptide|Gamma brain natriuretic peptide|Gamma-brain natriuretic peptide|Natriuretic peptide precursor B|NT proBNP
- ANFB_HUMAN|BNP|BNP(1-32)|BNP(5-29)|BNP-32|Brain type natriuretic peptide|Gamma brain natriuretic peptide|Gamma-brain natriuretic peptide|Natriuretic peptide precursor B|NT proBNP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16860
- P16860
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Natriuretic peptides B protein (27-102AA)
- Natriuretic peptides B protein (27-102AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together