Anti-NPEPL1 Rabbit Polyclonal Antibody
77744-771
New Product
- Antigen Symbol:NPEPL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q8NDH3
- Antigen Synonyms:Aminopeptidase like 1|Aminopeptidase-like 1|bA261P9.2|KIAA1974|NPEPL 1|NPEPL1|PEPL1_HUMAN|Probable aminopeptidase NPEPL1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Probable aminopeptidase NPEPL1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NPEPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat