Anti-LATS1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: LATS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-845
- 77742-000
- Antigen Symbol:
- LATS1
- Antigen Synonyms:
- h-warts|Large tumor suppressor homolog 1|LATS large tumor suppressor homolog 1|LATS1|LATS1_HUMAN|Serine threonine protein kinase LATS1|Serine/threonine-protein kinase LATS1|WARTS|WARTS protein kinase|wts
- h-warts|Large tumor suppressor homolog 1|LATS large tumor suppressor homolog 1|LATS1|LATS1_HUMAN|Serine threonine protein kinase LATS1|Serine/threonine-protein kinase LATS1|WARTS|WARTS protein kinase|wts
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O95835
- O95835
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase LATS1 protein (1-280AA)
- Recombinant Human Serine/threonine-protein kinase LATS1 protein (1-280AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together