Anti-FOXE3 Rabbit Polyclonal Antibody
Catalog # 77736-740
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FOXE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13461
- Antigen Synonyms:Drosphilia Forkhead Homolog Like 12|FKHL 12|FKHL12|Forkhead Box E3|Forkhead box protein E3|Forkhead Related Activator 8|Forkhead related protein FKHL12|Forkhead-related protein FKHL12|Forkhead-related transcription factor 8|Foxe3|FOXE3 forkhead box E3|FOXE3_HUMAN|FREAC 8|FREAC-8|FREAC8
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Drosphilia Forkhead Homolog Like 12
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: FOXE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat