Anti-FEZF1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FEZF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-064
- 77855-300
- Antigen Symbol:
- FEZF1
- FEZF1
- Antigen Synonyms:
- Zinc finger protein 312B|FEZF1_HUMAN|Fez family zinc finger protein 1|FEZ family zinc finger 1|FEZ|Zinc finger protein 312 like|fez like
- FEZ|FEZ family zinc finger 1|Fez family zinc finger protein 1|fez like|FEZF1_HUMAN|Zinc finger protein 312 like|Zinc finger protein 312B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A0PJY2
- A0PJY2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fez family zinc finger protein 1 protein (1-115AA)
- Recombinant Human Fez family zinc finger protein 1 protein (1-115AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together