Anti-POTEB Rabbit Polyclonal Antibody
Catalog # 77738-213
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:POTEB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q6S5H4
- Antigen Synonyms:POTEB|A26B1|POTE15|POTE ankyrin domain family member B|ANKRD26-like family B member 1|Prostate| ovary, testis-expressed protein on chromosome 15|POTE-15
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:POTE-15
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, ELISA:1:5000
Type:
Antigen: POTEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human