Anti-FGF13 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: FGF13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77855-332
- 77737-102
- Antigen Symbol:
- FGF13
- FGF13
- Antigen Synonyms:
- FGF 13|FGF 2|FGF-13|FGF13_HUMAN|FGF2|FHF 2|FHF-2|FHF2|Fibroblast growth factor 13|Fibroblast growth factor homologous factor 2|OTTHUMP00000024143|OTTHUMP00000024144
- FGF 13|FGF 2|FGF-13|FGF13_HUMAN|FGF2|FHF 2|FHF-2|FHF2|Fibroblast growth factor 13|Fibroblast growth factor homologous factor 2|OTTHUMP00000024143|OTTHUMP00000024144
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92913
- Q92913
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fibroblast growth factor 13 protein (1-245AA)
- Recombinant Human Fibroblast growth factor 13 protein (1-245AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together