Anti-GRK4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRK4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-468
- 77739-047
- Antigen Symbol:
- GRK4
- GRK4
- Antigen Synonyms:
- G protein coupled receptor kinase 4|G protein-coupled receptor kinase 2 (Drosophila)-like|G protein-coupled receptor kinase 4|G protein-coupled receptor kinase G proteinm coupled receptor kinase GPRK2L|GPRK4|GRK4_HUMAN|GRK4a|ITI1
- G protein coupled receptor kinase 4|G protein-coupled receptor kinase 2 (Drosophila)-like|G protein-coupled receptor kinase 4|G protein-coupled receptor kinase G proteinm coupled receptor kinase GPRK2L|GPRK4|GRK4_HUMAN|GRK4a|ITI1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32298
- P32298
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human G protein-coupled receptor kinase 4 protein (1-245AA)
- Recombinant Human G protein-coupled receptor kinase 4 protein (1-245AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together