Anti-IFNG Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFNG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-904
- 77740-700
- Antigen Symbol:
- IFNG
- IFNG
- Antigen Synonyms:
- IF 1|IFG|IFI|IFN gamma|IFN immune|IFN|immune|IFN-gamma|IFNG_HUMAN|Immune interferon|Interferon gamma|Type II Interferon
- IF 1|IFG|IFI|IFN gamma|IFN immune|IFN|immune|IFN-gamma|IFNG_HUMAN|Immune interferon|Interferon gamma|Type II Interferon
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01579
- P01579
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon gamma protein (24-162AA)
- Recombinant Human Interferon gamma protein (24-162AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together