Order Entry
United States
ContactUsLinkComponent
 
Anti-MXD3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-MXD3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77743-676
  • 77860-153
  • Antigen Symbol:
  • MXD3
  • MXD3
  • Antigen Synonyms:
  • bHLHc13|Class C basic helix-loop-helix protein 13|FLJ35523|Likely ortholog of mouse Max dimerization protein 3|MAD3_HUMAN|MAX dimerization protein 3|Max dimerizer 3|Max-associated protein 3|Max-interacting transcriptional repressor MAD3|MGC2383|Myx|OTTHUMP00000223596|OTTHUMP00000223598
  • bHLHc13|Class C basic helix-loop-helix protein 13|FLJ35523|Likely ortholog of mouse Max dimerization protein 3|MAD3_HUMAN|MAX dimerization protein 3|Max dimerizer 3|Max-associated protein 3|Max-interacting transcriptional repressor MAD3|MGC2383|Myx|OTTHUMP00000223596|OTTHUMP00000223598
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9BW11
  • Q9BW11
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Max dimerization protein 3 protein (1-206AA)
  • Recombinant Human Max dimerization protein 3 protein (1-206AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together