Anti-IGFBP1 Rabbit Polyclonal Antibody
Catalog # 77738-397
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IGFBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P08833
- Antigen Synonyms:AFBP|Alpha pregnancy associated endometrial globulin|Amniotic fluid binding protein|Binding protein 25|Binding protein 26|Binding protein 28|Growth hormone independent binding protein|hIGFBP 1|hIGFBP1|IBP 1|IBP-1|IBP1|IBP1_HUMAN|IGF binding protein 1|IGF BP25|IGF-binding protein 1|IGFBP 1|IGFBP-1|IGFBP1|Insulin like growth factor binding protein 1|Insulin-like growth factor-binding protein 1|Placental protein 12|PP 12|PP12
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:AFBP
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human