Anti-GMPPA Rabbit Polyclonal Antibody
Catalog # 77738-507
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GMPPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96IJ6
- Antigen Synonyms:GDP mannose pyrophosphorylase A|GDP-mannose pyrophosphorylase A|Gmppa|GMPPA_HUMAN|GTP mannose 1 phosphate guanylyltransferase alpha|GTP-mannose-1-phosphate guanylyltransferase alpha|Mannose 1 phosphate guanylyltransferase (GDP)|Mannose-1-phosphate guanyltransferase alpha
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Mannose-1-phosphate guanyltransferase alpha
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GMPPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat