Order Entry
United States
ContactUsLinkComponent
Anti-MEF2A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MEF2A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MEF2A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77742-981
  • 77859-641
  • Antigen Symbol:
  • MEF2A
  • MEF2A
  • Antigen Synonyms:
  • ADCAD1|MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A)|MEF2|MEF2A_HUMAN|Myocyte enhancer factor 2A|Myocyte-specific enhancer factor 2A|RSRFC4|RSRFC9|Serum response factor like protein 1|Serum response factor-like protein 1
  • ADCAD1|MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A)|MEF2|MEF2A_HUMAN|Myocyte enhancer factor 2A|Myocyte-specific enhancer factor 2A|RSRFC4|RSRFC9|Serum response factor like protein 1|Serum response factor-like protein 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q02078
  • Q02078
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant human Myocyte-specific enhancer factor 2A protein (72-226AA)
  • Recombinant human Myocyte-specific enhancer factor 2A protein (72-226AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together