Anti-NAPRT Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NAPRT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-913
- 77743-914
- Antigen Symbol:
- NAPRT
- NAPRT
- Antigen Synonyms:
- FHA HIT interacting protein|FHA-HIT-interacting protein|FHIP|NAPRT1|NAPRTase|Nicotinate phosphoribosyltransferase|Nicotinate phosphoribosyltransferase domain containing 1|Nicotinate phosphoribosyltransferase domain-containing protein 1|Nicotinic acid phosphoribosyltransferase|PNCB_HUMAN|PP3856
- FHA HIT interacting protein|FHA-HIT-interacting protein|FHIP|NAPRT1|NAPRTase|Nicotinate phosphoribosyltransferase|Nicotinate phosphoribosyltransferase domain containing 1|Nicotinate phosphoribosyltransferase domain-containing protein 1|Nicotinic acid phosphoribosyltransferase|PNCB_HUMAN|PP3856
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6XQN6
- Q6XQN6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nicotinate phosphoribosyltransferase protein (1-310AA)
- Nicotinate phosphoribosyltransferase protein (1-310AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together