Anti-Hspa5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Hspa5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77857-721
- 77740-477
- Antigen Symbol:
- Hspa5
- Hspa5
- Antigen Synonyms:
- Grp78|Endoplasmic reticulum chaperone BiP|EC 3.6.4.10|78 kDa glucose-regulated protein|GRP-78|Binding-immunoglobulin protein|BiP|Heat shock protein 70 family protein 5|HSP70 family protein 5|Heat shock protein family A member 5|Immunoglobulin heavy chain-binding protein
- Grp78|Endoplasmic reticulum chaperone BiP|EC 3.6.4.10|78 kDa glucose-regulated protein|GRP-78|Binding-immunoglobulin protein|BiP|Heat shock protein 70 family protein 5|HSP70 family protein 5|Heat shock protein family A member 5|Immunoglobulin heavy chain-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20029
- P20029
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant mouse Endoplasmic reticulum chaperone BiP protein (20-655AA)
- Recombinant mouse Endoplasmic reticulum chaperone BiP protein (20-655AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together