Anti-PFDN5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PFDN5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-171
- 77859-790
- Antigen Symbol:
- PFDN5
- Antigen Synonyms:
- 1190001O17Rik|1700010A06Rik|c myc binding protein|c-myc binding protein MM 1|C-myc-binding protein Mm-1|D15Ertd697e|EIG 1|Eig1|MGC5329|MGC71907|MM 1|MM1|Myc modulator 1|PFD5|PFD5_HUMAN|PFDN5|Prefoldin 5|Prefoldin subunit 5
- 1190001O17Rik|1700010A06Rik|c myc binding protein|c-myc binding protein MM 1|C-myc-binding protein Mm-1|D15Ertd697e|EIG 1|Eig1|MGC5329|MGC71907|MM 1|MM1|Myc modulator 1|PFD5|PFD5_HUMAN|PFDN5|Prefoldin 5|Prefoldin subunit 5
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q99471
- Q99471
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Prefoldin subunit 5 protein (2-154AA)
- Recombinant Human Prefoldin subunit 5 protein (2-154AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together